Coulomat oils for Karl Fischer titration, HYDRANAL™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
34868-500ML 34868-100ML 34868-6X100ML 34868-6X500ML
34868-500MLEA 134 EUR
34868-500ML 34868-100ML 34868-6X100ML 34868-6X500ML
Coulomat oils for Karl Fischer titration, HYDRANAL™
Karl Fischer titranten en reagentia
Coulomat oil is a reagent for coulometric KF titration in oils (anolyte solution), preferred to use for the cells with diaphragm.