Natriumhydroxide 0,01 mol/l (0,01 N) Volumetric
Leverancier: PanReac AppliChem
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Caustic soda
181845.1211 181845.1210
APPC181845.1211EA 35.3 EUR
APPC181845.1211 APPC181845.1210
Natriumhydroxide 0,01 mol/l (0,01 N) Volumetric
Natriumhydroxide
Formule: NaOH
MW: 40 g/mol
Kookpunt: 1390
Smeltpunt: 323
MDL Nummer: MFCD00003548
CAS nummer: 1310-73-2
UN: 1824
ADR: 8 III
Merck Index: 14,08627