Hank's balanced salts, MP Biomedicals
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
091710120 091710122
ICNA091710120EA 50.1 EUR
ICNA091710120 ICNA091710122
Hank's balanced salts, MP Biomedicals
Buffers Buffers voor weefselkweek
Hanks' balanced salts solution is used to maintain the pH and osmotic balance in the medium and to provide the cells with water and essential inorganic ions.