Cell culture media, powdered, Murashige and Skoog medium, MP Biomedicals
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
092610020 092610022 092610024
ICNA092610020EA 64.6 EUR
ICNA092610020 ICNA092610022 ICNA092610024
Cell culture media, powdered, Murashige and Skoog medium, MP Biomedicals
Celcultuurmedia
Murashige and Skoog Medium is widely used plant tissue culture growth medium. It has proven effective in the culture of tissue derived from monocotyledons and dicotyledons.
This medium, originally formulated to support tobacco callus, has been used to support callus initiation and growth, the growth of cells in suspension culture and the regeneration of shoots and plantlets from explants. More recently, it has been used for micropropagation of ornamental, vegetable and fruit plants in research and commercial enterprises.
The medium contains Murashige and Skoog salts, as well as a mixture of vitamins. This modification of the original formula does not contain agar, sucrose, IAA, and kinetin.