Transformation kit, EZ-Yeast™
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
EZ Yeast™
ICNA112100200EA 103 EUR
ICNA112100200
Transformation kit, EZ-Yeast™
Reagentia voor nucleïnezuren Reagentia voor transformatie
The EZ-Yeast™ Transformation Kit is designed for high throughput transformation with efficiencies up to 10³ transformants per μg plasmid DNA. It is ideal for cases where a large number of transformations must be performed, but only a few transformants are needed. Perfect for a quick analysis of potentially positive two-hybrid colonies, it does not require fully competent cells prior to transformation.
Leveringsinformatie: Kit contains EZ-Transformation Solution and carrier DNA.