Binocular telescopes, teleMED
Leverancier: ESCHENBACH OPTIK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1634 16344
ESCH1634EA 404 EUR
ESCH1634 ESCH16344
Binocular telescopes, teleMED
Vergrootglazen en loepen
These telescopes are available in a binocular system, offering a bright image, with a plus objective lens and a minus ocular lens.
  • Sturdy frame made from glass fibre reinforced black plastic
  • Achromatic lens made of high quality silicate
  • Adjustable pupillary distance from 54 to 74 mm
  • Focusing dioptre compensation for each eyepiece
  • Comfortable and easily adjustable saddle bridge
Leveringsinformatie: Supplied in a black zippered case.