Water, Molecular biology grade_meetSpec_USP Molecular biology grade, sterile, Corning®
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Hydrogen oxide
46-000-CM 46-000-CI 46-000-CV
46-000-CMEA 191.38 EUR
46-000-CM MDTC46-000-CI MDTC46-000-CV
Water, Molecular biology grade_meetSpec_USP Molecular biology grade, sterile, Corning®
Water
Molecular biology grade water is ideal for a wide variety of routine tasks requiring a high degree of sterility Molecular biology grade water is ideal for the preparation of reagents, rinsing glassware and plasticware, and other molecular biology applications where RNase-, DNase-, and Protease-free water is required. No toxic agents, including DEPC, are used in the preparation of Mediatech's molecular biology grade water, eliminating possible interferences with enzymatic reactions.
Certificaten: Molecular Biology Grade Water Tested to USP Sterile Purified Water Specifications
Formule: H₂O
MW: 18.02 g/mol
Kookpunt: 100
Smeltpunt: 0
MDL Nummer: MFCD00011332
CAS nummer: 7732-18-5
Merck Index: 13,10098