VWR® Hichrom Organic Acid, HPLC Columns
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HICH9046EA 2040 EUR
HICH9046 HICH9048 HICH9646 HICH9648 HICH28883 HICH28884
VWR® Hichrom Organic Acid, HPLC Columns
Kolommen voor chromatografie
Avantor® Hichrom Organic Acid columns are ion exclusion columns packed with sulphonated polystyrene-divinylbenzene.
  • Rapid analysis of organic acids and alcohols
  • pH stable polymer resin
  • Isocratic 100% aquoeus mobile phases only, no organic solvents
  • USP L17
The OA-1000 and OA-2000 columns exhibit excellent selectivity for aliphatic and aromatic acids. As with most ion exclusion columns, a column heater is necessary for normal operating procedures. The IOA-1000 and IOA-2000 columns are suitable for the separation of citric and other acids from glucose and fructose.