ESD safety shoes, sandals, Velcro® fastening, Laslo
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
716271/38 716271/39 716271/40 716271/41 716271/42 716271/43 716271/44 716271/45 716271/46 716271/47 716271/48 2716271/49 2716271/50
114-8046EA 81.9 EUR
114-8046 114-8047 114-8048 114-8049 114-8050 114-8051 114-8052 114-8053 114-8054 114-8055 114-8056 114-8057 114-8058
ESD safety shoes, sandals, Velcro® fastening, Laslo
Schoenen en laarzen
Lightweight ESD safety sandal made from breathable cowhide leather, textile materials and microfibre, a padded tongue and collar plus a slip resistant, dual-density PU/PU sole (anti-static, oil and fuel resistant also heat resistant up to 130 °C).
  • Climate-regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full length fleece inlay sole (ESD Pro), black
  • Integrated heel loop
  • Metal-free puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in dry work areas such as industry, storage, transport, or assembly and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
Certificaten: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S1P, SRC.