Chemisch bestendige handschoenen, Alto 285
Leverancier: MAPA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
34285019 34285360 34285018
112-5550EA 37.1 EUR
112-5550 112-5548 112-5549
Chemisch bestendige handschoenen, Alto 285
Handschoenen
Liquid-proof glove made of natural rubber latex. Curved fingers and contoured palm. Smooth chlorinated internal surface. Non-slip finish in hand area. Guaranteed silicone-free.
  • Raised pattern on hand area
  • Extended cuff to protect the whole arm
  • Easy to put on, to take off and to clean thanks to the chlorinated surface
  • Can be used with knitted underglove
  • Good resistance to mechanical hazard
  • Conforms to the FDA regulation for food contact
Designed for protection against chemicals such as acids, bases, detergents, alcohols, ketonic solvents. Not recommended for contact with petroleum, aromatic or chlorinated solvents.
Thickness: 1,0 mm
Colour: Tan
EN 374 (ABKL), EN 388 (2131), Cat. III
Certificaten: EN ISO 374-1 Type A (ABKMPT), EN ISO 374-5, EN 388:2016 (2131X), Cat. III.
Food contact approved.