Avantor® Apollo C8, HPLC Columns
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HICH36506EA 715 EUR
HICH36506 HICH36512 HICH5126774 96463/N
Avantor® Apollo C8, HPLC Columns
Kolommen voor chromatografie
Avantor® Apollo HPLC columns use high purity, base deactivated silica for powerful separations.
  • Easy scale-up from analytical to preparative
  • Extended pH stability is about 1,5 to 10,5
  • USP L7
These columns are ideal for routine analysis and for educational laboratories.