Dehydrated culture media, LB-medium, molecular biology grade, MP Biomedicals
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
113002095 113002075 113002085 113002065 113002265 113002275
ICNA113002095EA 501 EUR
ICNA113002095 ICNA113002075 ICNA113002085 ICNA113002065 ICNA113002265 ICNA113002275
Dehydrated culture media, LB-medium, molecular biology grade, MP Biomedicals
Media voor microbiologie Geprepareerde media voor microbiologie
LB Medium is a nutritionally rich medium originally developed for growth and maintenance of E. coli strains. It supports a luxurious growth of E. coli cells as tryptone and yeast extract supply essential growth factors that E. coli cells would otherwise have to synthesise. The medium also contains essential electrolytes for transport and osmotic balance, due to the NaCl component.
  • Available in pack pouches or in a case
  • Ready-to-use
  • LB Lennox media a molecular biology certified bacterial growth
  • Proprietary media for enhanced cell yield
  • Convenient packaging formats for a variety of applications and budgets
  • Proven and reliable reputation
  • High-quality raw materials and additives
Rich medium for the growth of E. coli and certified for plasmid and other bacterial cultures in liquid medium.
Application includes: Bacteria cell culture, gene expression, transformation, microbiological analysis, protein expression.