PCR master mix, Taq-&LOAD™, MP Biomedicals
Leverancier: MP Biomedicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
11EPTAL110
11EPTAL100
ICNA11EPTAL110EA
926
EUR
ICNA11EPTAL110
ICNA11EPTAL100
PCR master mix, Taq-&LOAD™, MP Biomedicals
Reagentia voor nucleïnezuren
End-point PCR Enzymes and Kits
Taq-&LOAD™ PCR Master Mix is an optimised 5X reaction mix for PCR containing recombinant Taq DNA polymerase, dNTPs, MgCl₂, and incubation and loading buffers. Also included are a compound increasing the density of the sample and a red/purple dye.
- Significant savings in preparation time
- Reduced pipetting
- Reduced risk of errors and contamination
- Stable at 4 °C
This mastermix allows direct loading onto the agarose gel after PCR without the need for additional loading dye. The included red purple dye migrates at 400 bp on a 1,2% agarose gel.