Taq DNA polymerase, MP Biomedicals
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
11EPTQD025 11EPTQD325 11EPTQC200 11EPTQD925 11EPTQA105 11EPTQA925 11EPTQA100 11EPTQA025 11EPTQA325 11EPTQC060 11EPTQL100
ICNA11EPTQD025EA 163 EUR
ICNA11EPTQD025 ICNA11EPTQD325 ICNA11EPTQC200 ICNA11EPTQD925 ICNA11EPTQA105 ICNA11EPTQA925 ICNA11EPTQA100 ICNA11EPTQA025 ICNA11EPTQA325 ICNA11EPTQC060 ICNA11EPTQL100
Taq DNA polymerase, MP Biomedicals
Reagentia voor nucleïnezuren End-point PCR Enzymes and Kits
Taq DNA Polymerase is a specialised thermostable enzyme isolated from the thermophilic bacterium Thermus aquaticus. The recombinant form of this enzyme is expressed in E. coli. It is highly processive, recombinant polymerase which catalyses the addition of mononucleotide units to the 3’-OH end of a primer chain. It remains functional even after prolonged incubation at 95 °C. This enzyme possesses 5’-3’ exonuclease activiy.
Taq DNA Polymerase is used for the DNA polymerase chain reaction (PCR) in order to amplify DNA sequences. Other applications include RT-PCR, DNA sequencing and DNA labeling.