Lysing kits for Fastprep®-24 5G, Lysing Matrix F, MP Biomedicals
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
116915050 116915100 116915500 116935025
ICNA116915050EA 225 EUR
ICNA116915050 ICNA116915100 ICNA116915500 ICNA116935025
Lysing kits for Fastprep®-24 5G, Lysing Matrix F, MP Biomedicals
Homogenisatoren Tafelmodel homogenisatoren
Each impact-resistant tube contains 1,6 mm aluminum oxide particles and 1,6 mm silicon carbide particles.
  • Effective for applications such as RNA and protein isolation
  • Irregular, angular shape of the crystals give lower shearing
  • Matrix F tubes have white caps
  • Can be stored at room temperature
  • DNase and RNase free
Silicon carbide has relatively low density and high strength and hardness, and therefore is effective at breaking tough, hard, or brittle cell membranes. This is useful in applications such as isolation of intact organelles, and super-molecular structures where lysis conditions necessitate high impaction but low shearing.