Lysing kits for Fastprep®-24 5G, Lysing Matrix I, MP Biomedicals
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
116918050 116918100
ICNA116918050EA 225 EUR
ICNA116918050 ICNA116918100
Lysing kits for Fastprep®-24 5G, Lysing Matrix I, MP Biomedicals
Homogenisatoren Tafelmodel homogenisatoren
Each impact-resistant 2 ml tube contains 2 mm yellow zirconia beads and a 4 mm black ceramic sphere.
  • Can be used for dry grinding fungal spores to isolate RNA and proteins
  • Matrix I tubes have clear caps
  • Can be stored at room temperature (15 to 30 °C)
  • DNase and RNase free
Zirconium oxide has very high density and high tensile strength and hardness, and is effective at breaking very tough and hard samples, including organisms within a tough or dense exterior matrix. This is useful in applications such as isolation of intact organelles, and super-More Product Information