Lysing kits for Fastprep®-24 5G, Lysing Matrix J, MP Biomedicals
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
116919050 116919100
ICNA116919050EA 225 EUR
ICNA116919050 ICNA116919100
Lysing kits for Fastprep®-24 5G, Lysing Matrix J, MP Biomedicals
Homogenisatoren Tafelmodel homogenisatoren
Each impact-resistant 2 ml tube contains 2 mm yellow zirconia beads and 1,6 mm aluminum oxide particles.
  • Matrix J tubes have pink caps
  • Can be store at room temperature
  • DNase and RNase Free
Zirconium oxide has very high density and high tensile strength and hardness, and is effective at breaking very tough and hard samples, including organisms within a tough or dense exterior matrix. This is useful in applications such as isolation of intact organelles, and super-molecular structures where lysis conditions necessitate high impaction but low shearing.More Product Information