Lysing kits for Fastprep®-24 5G, Lysing Matrix SS, MP Biomedicals
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
116540431 116921100
ICNA116540431EA 181 EUR
ICNA116540431 ICNA116921100
Lysing kits for Fastprep®-24 5G, Lysing Matrix SS, MP Biomedicals
Homogenisatoren Tafelmodel homogenisatoren
Each impact-resistant 2 ml tube contains one 6,35 mm, 0,25 cal stainless-steel grinding ball.
  • Can be stored at room temperature
  • DNase and RNase free
Use to break hard/brittle samples. Specialty applications include RNA extraction from difficult samples where lysis by impaction only is preferable with minimal amounts of mechanical shearing.