Grinding matrix for lysing kits, MP Biomedicals
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
116540424 116040801 116540034 116540412 116540422
ICNA116540424EA 122 EUR
ICNA116540424 ICNA116040801 ICNA116540034 ICNA116540412 ICNA116540422
Grinding matrix for lysing kits, MP Biomedicals
Parels Keramische kralen
Ceramic sphere is a Zirconium oxide coated grinding matrix which is chemically inert and will not bind nucleic acids. These are components of lysing matrices.