Lysing kits for Fastprep®-24 5G, Lysing Matrix S, MP Biomedicals
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
116925100 116925500 116925050 116925000
ICNA116925100EA 357 EUR
ICNA116925100 ICNA116925500 ICNA116925050 ICNA116925000
Lysing kits for Fastprep®-24 5G, Lysing Matrix S, MP Biomedicals
Homogenisatoren Tafelmodel homogenisatoren
Metal bead lysing matrix contains 3,175 mm diameter stainless steel.
  • Use to break hard/brittle samples
  • Can be stored at room temperature
  • DNase and RNase free
Specialty applications include RNA and protein extraction from difficult samples where lysis by impaction only is preferable with minimal amounts of mechanical shearing.