Lysing kits for Fastprep®-24 5G, Lysing Matrix Y, MP Biomedicals
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
116960500 116960100 116960050 116975005
ICNA116960500EA 1429 EUR
ICNA116960500 ICNA116960100 ICNA116960050 ICNA116975005
Lysing kits for Fastprep®-24 5G, Lysing Matrix Y, MP Biomedicals
Homogenisatoren Tafelmodel homogenisatoren
Each impact-resistant 2 ml tube contains 0,5 mm diameter yttria-stabilised zirconium oxide beads.
  • Matrix tubes contain white caps
  • Can be stored at room temperature (15 to 30 °C)
  • DNase and RNase free
Lysing Matrix Y is used primarily for lysis of gram positive and gram negative bacteria, fungal tissue and spores, yeasts, moulds, filamentous algaes and biofilms. Y₂O₃ and ZrO₂ have high density and extremely high hardness making this matrix ideal for cracking of very tough cell walls, while it's smooth surface minimises shearing.
Order Now
Specificaties
Environmentally Preferable