Chemical resistant overalls, Tritex® pro
Leverancier: Zellstoff-Vertriebs-GmbH & Co. KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
30623-90 30624-90 30625-90 30626-90
113-7871EA 209 EUR
113-7871 113-7872 113-7873 113-7874
Chemical resistant overalls, Tritex® pro
Wegwerpkleding
Tritex® pro protective clothing is dust-tight, chemical resistant and partially splashproof and is made using an innovative in-line procedure. These extremely robust overalls are made from 3-ply SMS material (spunbond-meltblown-spunbond). A meltblown filter membrane is sandwiched between two durable outer layers.
  • Provide reliable protection against fine dust and splashes from liquid chemicals
  • Excellent breathability: Filter membrane prevents excess heat or moisture, even when worn for a long time
  • Elasticated hood, zip cover strip, elasticated waist and elasticated arms and legs
Extremely versatile; applications include inspection, maintenance and decontamination work, handling material containing fibreglass and asbestos removal.
Colour: Blue
Certificaten: CE 0624, Cat. III, Type 5 + 6