Beschermende overall, 4520
Leverancier: 3M
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4520L 4520M 4520XL 4520XXL 4520S 4520CSL 4520CSXL 4520CSM 4520CSXXL
113-0795EA 230 EUR
113-0795 113-0796 113-0797 113-0798 MMMA4520S MMMA4520CSL MMMA4520CSXL MMMA4520CSM MMMA4520CSXXL
Beschermende overall, 4520
Wegwerpkleding
3M™ Beschermende Overall 4520 is ontworpen om meerdere lagen SMMMS te bieden voor effectieve stevigheid en sterkte bij een lichtgewicht van 43 g/m². Uitstekend ademend en zacht met een stofachtig gevoel, het kan langdurig comfortabel worden gedragen en beschermt tegen statische ontlading, gevaarlijke deeltjes en vloeistofspatten.
  • Coveralls protect against certain light liquid splashes and hazardous dusts
  • Anti-static treatment can help reduce static build-up
  • Does not contain natural rubber latex components
  • Seamless shoulders and sleeve tops offer extra strength and protection
  • Extra-roomy design in arms and legs deliver enhanced mobility
  • Breathable material technology for reduced heat stress and premium wearer comfort all day
  • Knitted cuffs and elasticated waist and ankles for increased comfort and freedom of movement
  • Available in collared and hooded profiles, in a wide range of sizes
Use 3M™ Protective Coverall 4520 for protection against dirt, splashes, and toxic dusts, including nuclear particles. It meets a variety of standards for chemical protective clothing including EN 1149 Antistatic protection, EN 1073-2 Nuclear particulate resistance, EN ISO 13688, Type 6 Liquid Splash EN 13034, and Type 5 Dust EN ISO 13982-1. These garments are easily identifiable with the green triangle, which is made of the same 43 gsm SMMMS material.
This coverall has been engineered to reduce the weight and retain the performance through the addition of multiple layers. It’s made with SMMMS, which is an inner and outer layer of spunbond and 3 layers of meltblown material in the middle. The Spunbond acts as the smooth strength and splash protection, while the Meltblown layers provide an intricate multi-layered web of microfibres that filter out dry particulates. Soft, breathable, and very lightweight, the non-woven material allows for easier release of body heat and moisture, giving extended comfort wear for increased productivity in the workplace.
The 3-panel hood works well with other PPE, soft knitted cuffs offer secure and comfortable arm placement, and a 2-way zip ensures better fit and peace of mind. Offering antistatic performance to EN 1149 to minimise discharge and risk of sparks and ignition. Ideal for particulate based applications such as pharmaceutical processing and the handling of toxic powders.
Certificaten: CE Category III. EN 1073-2 Particulate Radioactive Contamination. EN 1149-5 Electrostatic Properties. EN 13034:2005+A1:2009 Type 6 Liquid Splash. EN ISO 13982-1:2004 Type 5 Hazardous Dusts.