EasySat™ bucketless floor mop
Leverancier: CONTEC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BLVKMOP BLVKMOPIR TBLVKMOP BLMOPQC
129-0213EA 571 EUR
129-0213 129-0214 129-0215 141-1467
EasySat™ bucketless floor mop
Moppen
EasySat™ bucketless floor mop is an effective and quick solution to cleaning challenges.
  • Fully autoclavable
  • Closed solution tank minimises exposure to chemicals
  • Large opening makes tank easy to fill
  • Lightweight mop system weighs 3,2 kg with full tank
  • Trigger flow control mechanism releases cleaning solution on demand or continuously
  • Angled mop head cleans hard-to-reach areas and is available in two styles: smooth surface for standard floor cleaning, and textured surface for more abrasive action
Designed for the cleanroom, this innovative mop is easy to use. The built-in 1,5 litre solution tank completely eliminates the need for a bucket and wringer (one full tank cleans up to 465 m²). Disposable mop heads are made from an ultra clean polyester foam VertiKlean.
Mop head is available validated sterile as per AAMI/ISO 11137:2006 guidelines. Gamma irradiated to SAL 10⁻⁶, one of the strictest guidelines established by AAMI/ISO.