VWR® Sealpette™ Pro Enkelkanaalspipetten, Mechanisch, Variabel Volume
Leverancier: JENCONS SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
480-044 480-039 480-097 480-098
613-1244EA 0 EUR
613-1244 613-0919 613-0922 613-0923
VWR® Sealpette™ Pro Enkelkanaalspipetten, Mechanisch, Variabel Volume
Pipetten Micropipetten
Robust, lightweight pipettes for both left- and right-handed users, featuring very low tip ejection and pipetting forces.
  • Thermal insulation of internal components
  • Fast and light volume adjustment with small increments and volume locking button
  • Clear and easy to read display
  • Personal identification with an ID tag
  • Easy maintenance with one multi-purpose tool (calibration, opening tool for ID window or tubes)
  • Fully autoclavable
The Sealpette™ tip cone is supplied with a Standard filter for protection against pipette contamination by aerosol or over-pipetting. There are two types of replacement filter available; The Standard filter for general applications and the Plus filter for more demanding applications such as molecular biology, cell culture, bacterial and virological work.
Bestelinformatie: Supplied with Standard filter, calibration tool and QC-certificate.