Screw caps with aperture
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
01321018 01321014 01321025 01321032
201-0247EA 2.6 EUR
201-0247 201-0246 201-0248 201-0249
Screw caps with aperture
Sluitingen Reageerbuissluitingen
PBT, red without seals.
  • For DIN GL threads
  • Heat resistant up to 180 °C
  • Autoclavable, sterilisable with hot air
With central hole.