Chemisch en snijbestendige handschoenen, Showa 3416
Leverancier: Showa
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3416-08 3416-10 3416-11 3416-09
112-5784EA 278 EUR
112-5784 112-5786 112-5787 112-5785
Chemisch en snijbestendige handschoenen, Showa 3416
Handschoenen
Constructed with a seamless knit liner for maximum comfort and endurance, this fully coated neoprene gauntlet provides ANSI level A4 and EN level 5 cut protection and protection against acids, caustics, solvents, grease and oil.
  • Seamless liner made from cut resistant engineered fibre
  • Fully coated
  • Flexible coating with maximum comfort and protection
  • Rough particle finish
  • Gauntlet cuff for protection of the forearm
Recommended industries: Transport, steel erecting, mechanical, metallurgy, petrochemical, automotive.
Certificaten: EN 388:2016 (3X43E), EN ISO 374-1 Type A (AJKLMT), EN ISO 374-5; Cat. III