Safety boots, pull on, Jalfrigg
Leverancier: Jallatte
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
J0652/35 J0652/36 J0652/37 J0652/38 J0652/39 J0652/40 J0652/41 J0652/42 J0652/43 J0652/44 J0652/45 J0652/46 J0652/47 J0652/48
114-4013EA 184 EUR
114-4013 114-4014 114-4015 114-4016 114-4017 114-4018 114-4019 114-4020 114-4021 114-4022 114-4023 114-4024 114-4025 114-4026
Safety boots, pull on, Jalfrigg
Schoenen en laarzen
Metal-free safety boots in natural full-grain leather, highly water and oil resistant.
  • Softane™ dual-density PU provides excellent absorption of heel shocks and vibrations
  • Lightweight, non magnetic, perforated composite toecap (200 J)
  • Non magnetic, lightweight and flexible FleXtane™ HP midsole (100% composite)
Fully lined for use in cold environments.
Colour: Beige
Certificaten: EN ISO 20345; S3, CI, SRC