One-piece fingertight fittings
Leverancier: VICI JOUR RESEARCH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
JR-5502-5 JR-5507-5 JR-5508-5
554-1513EA 108 EUR
554-1513 554-1515 554-1516
One-piece fingertight fittings
Fittingen en connectoren
PEEK one-piece fingertight fittings. 10-32 threads, for 1/16" tubing. Fingertight up to 350 bar (5000 psi).
  • Excellent chemical resistance
  • Machined, not molded
  • One-piece – no ferrule required
  • Biocompatible