Staining dishes
Leverancier: BIO OPTICA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
10-30 10-10 10-20 10-33
BOPT10-30EA 136 EUR
BOPT10-30 BOPT10-10 BOPT10-20 BOPT10-33
Staining dishes
Potjes Kleurbakjes
These staining dishes are made of PET (polyethylene terephthalate) stiffened with glass fibres. They are resistant against solvents especially in cytology and histology staining processes.
  • Resist temperature between 0 to 170 °C
  • Can be used with immunohistochemistry procedures