Empore™ Extraction Disks, SPE Disk
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Empore®
SIAL66887-UEA 187 EUR
SIAL66887-U SIAL66898-U SIAL66888-U 66884-U SIAL66896-U SIAL66894-U 66883-U
Empore™ Extraction Disks, SPE Disk
Filters Filterschijven
Designed for extracting large volumes of aqueous samples. Suitable for EPA methods and environmental methods.
  • Provides an efficient alternative to liquid-liquid extraction (LLE)
  • Developed for efficient extraction of pollutants