Standard hydrometers, M100
Leverancier: Alla France
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3102MC06060BQP 3102MC07060BQP 3102MC08060BQP 3102MC09060BQP 3102MC10060MQP 3102MC11060MQP 3102MC12060MQP 3102MC14060MQP 3102MC15060MQP 3102MC16060MQP 3102MC17060MQP
ALLA3102MC06060BQPEA 25.1 EUR
ALLA3102MC06060BQP ALLA3102MC07060BQP ALLA3102MC08060BQP ALLA3102MC09060BQP 614-1634 614-1635 614-1636 614-1638 614-1639 614-1640 614-1641
Standard hydrometers, M100
Hydrometers
These specific gravity hydrometers are marked with reference norms, calibration temperature, surface tension, individual and serial numbers.
  • Reading below the meniscus
  • Calibration temperature of 15 °C
  • Benchmark on the stem to show any movement of the scale
Calibrated according to the following tensions:
0,600 to 1,000 Sp.Gr.= 16 to 35 mN/m (low tension)
1,000 to 1,300 Sp.Gr.= 55 mN/m (medium tension)
Certificaten: ISO 650.
Bestelinformatie: 'EU' Traceable Calibration Certificate or 'EO' Official Calibration Certificate is available on request.More Product Information