Sulphide reagents 1 and 2
Leverancier: Hach
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
181632 181732 181749
HACH181632EA 33 EUR
HACH181632 HACH181732 HACH181749
Sulphide reagents 1 and 2
Milieutestkits
Used for sulphide determination by the methylene blue method.
  • Range upto 800 µg/L
  • Used with visual test kits