EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) 98, Laboratory Reagent, J.T.Baker®
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: EGTA, Ethylenebis(oxyethylenenitrilo)tetra(acetic acid), Kestranal® FE PA, Idranal® VI, Ethylene glycol-O,O'-bis(2-aminoethyl)-N,N,N',N'-tetraacetic acid, Ethylenebis(oxyethylenenitrilo)tetraacetic acid, Ethylene Glycol Bis(2-aminoethyl Ether)-N,N,N,N-tetraacetic acid, 3,12-Bis(carboxymethyl)-6,9-dioxa-3,12-diazatetradecanedioic acid, Titriplex® VI
M.O.C. Category N
L657-06EA 0 EUR
L657-06 L657-05
EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) 98, Laboratory Reagent, J.T.Baker®
EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid)
Formule: C₁₄H₂₄N₂O₁₀
MW: 380.35 g/mol
Smeltpunt: 240…244 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004291
CAS nummer: 67-42-5
EINECS: 200-651-2
Merck Index: 13,03560
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use = 98 %
Assay (C₁₄H₂₄N₂O₁₀) ≥98 %
Specificaties
Environmentally Preferable