Zoutzuur Ph. Eur., Macron Fine Chemicals™
Leverancier: Avantor Performance Materials
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Hydrochloric acid solution, Muriatic acid
P112-33EA 30.79 EUR
P112-33 P112-36
Zoutzuur Ph. Eur., Macron Fine Chemicals™
Zoutzuur
Formule: HCl
MW: 36.46 g/mol
MDL Nummer: MFCD00011324
CAS nummer: 7647-01-0
UN: 1789
ADR: 8 II