Petroleum spirit, 40…60 °C, AR®, Laboratory Reagent, Macron Fine Chemicals™
Leverancier: Avantor Performance Materials
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Gasoline, Petroleum ether, Petrol, Benzine
6722-45EA 546.03 EUR
6722-45 6722-25
Petroleum spirit, 40…60 °C, AR®, Laboratory Reagent, Macron Fine Chemicals™
Petroleum spirit
Dichtheid: 0.7
Vlampunt: -10
MDL Nummer: MFCD00081849
CAS nummer: 64742-49-0
UN: 1268
ADR: 3 II