Chairs, Semi Stand-Up and Stools, V - KORR and 316 L Stainless Steel
Leverancier: Kango
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3VN36VHXX00903 3VN36VHXX04903 3VN12VHXX00903 3VN26VHXX00903 3VN12VHXX04903 3VN26VHXX04903
BIEN3VN36VHXX00903EA 2160 EUR
BIEN3VN36VHXX00903 BIEN3VN36VHXX04903 BIEN3VN12VHXX00903 BIEN3VN26VHXX00903 BIEN3VN12VHXX04903 BIEN3VN26VHXX04903
Chairs, Semi Stand-Up and Stools, V - KORR and 316 L Stainless Steel
Stoelen
Chairs, stools and stand-up with resistant V-KORR seats and backrests with stainless steel 316L base. Ideal for use in wet rooms, cleanrooms, laboratories and in the food industry.
  • 316L stainless steel column and base, resistant to corrosion
  • Can be washed with hosepipes
  • Autoclavable
  • Height adjustment with screw hidden inside the column
  • Chemical resistance in « litterature »
Chairs:
Ergonomically designed seats and backrests
Available with or without footrest
Dimensions of the seat (W×D):410×390 mmDimensions of the backrest (W×H): 400×220 mm
Stools:
Round seat
Available with or without footrest
Diameter of the seat: 310 mm