Vacuum adapter, ground joint, PTFE, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
N1655-04 N1655-06
BOHLN1655-04EA 84.1 EUR
BOHLN1655-04 BOHLN1655-06
Vacuum adapter, ground joint, PTFE, BOLA
Adapters Adapters, Slijpstukken met slangaansluitingen
This vacuum adapter is made of PTFE with O-ring made of FKM for secure placement of standard filter cartridges made of glass or plastic on vessels and round flasks with ground joint.
  • Lateral step-type solenoid made of PTFE
  • Chemical resistance
  • Temperature resistance from –15 to +200 °C
For vacuum filtration, the O-ring at the passageway seals the connection to the filter slide so that even a small vacuum is sufficient.
Order Now
Specificaties
Environmentally Preferable