Solvent desorption tubes, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
21014 21026-U 21024-U 505293 505315 505320-U 505323 505331 505358
21014.EA 798 EUR
21014. 21026-U 21024-U 505293. 505315. 505320-U 505323. 505331. 505358.
Solvent desorption tubes, Supelco®
Kolommen voor chromatografie
These LpDNPH cartridges are suitable for sampling carbonyls in ambient, indoor, and industrial atmospheres. The derivatives are eluted from the cartridge in acetonitrile and are analysed by HPLC.
The S10x cartridge fits automated samplers. The S10L cartridge is for analysts who prefer the shorter dimensions and do not need an adapter for sampling. H10, H30, and H300 cartridges contain higher loadings of 2,4-DNPH, making them suitable for use in high concentration environments. The disposable ozone scrubber is similar to an S10L cartridge, containing 1,5 g of high purity potassium iodide.