Test kits, photometric, Phenols, LCK cuvette tests
Leverancier: Hach
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
LCK345
LCK346
705-3057EA
150
EUR
705-3057
705-3058
Test kits, photometric, Phenols, LCK cuvette tests
Milieutestkits
Ready for use, photometric evaluation system and environmentally friendly.
- Precise and reliable measurement values
- Maximum safety for users, thanks to the closed system and low amounts of reagents
- Convenient and error-free dosing of the reagents
- Barcode label for automatic recognition in the photometer
- Differentiation between tests and measuring ranges by means of colour-coding
Storage conditions: 15 to 25 °C for LCK345; 2 to 8 °C (keep refrigerated) for LCK346
Certificaten: LCK346 according to standard ISO 6439-1990, DIN 38409 H16.