Test kits, photometric, Sulphate, LCK cuvette tests
Leverancier: Hach
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
LCK153 LCK353
705-3066EA 164 EUR
705-3066 705-3067
Test kits, photometric, Sulphate, LCK cuvette tests
Milieutestkits
Pre-dosed reagents for maximum safety.
  • Precise and reliable measurement values
  • Maximum safety for users, thanks to the closed system and low amounts of reagents
  • Convenient and error-free dosing of the reagents
  • Barcode label for automatic recognition in the photometer
  • Differentiation between tests and measuring ranges by means of colour-coding
Storage conditions: 15 to 25 °C