Test kits, photometric, TOC, LCK cuvette tests
Leverancier: Hach
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
LCK380 LCK381 LCK385 LCK386 LCK387
705-3024EA 256 EUR
705-3024 705-3025 705-3021 705-3022 705-3023
Test kits, photometric, TOC, LCK cuvette tests
Milieutestkits
Pre-dosed reagents for maximum safety.
  • Precise and reliable measurement values
  • Maximum safety for users, thanks to the closed system and low amounts of reagents
  • Convenient, error-free dosing of the reagents
  • Barcode label for automatic recognition in the photometer
  • Differentiation between tests and measuring ranges by means of colour coding
Storage conditions: 15 - 25 °C
Certificaten: DIN 38409-H3
Order Now
Specificaties
Environmentally Preferable