Craft knife handles, No. 5A / No. 6A
Leverancier: SWANN MORTON
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1006 1005
SWAN1006EA 273 EUR
SWAN1006 SWAN1005
Craft knife handles, No. 5A / No. 6A
Scalpels Scalpelhandvaten
These acrylic craft knife handles have a stainless blade fitting.
Verpakking: Individually packed without blades.