Craft knife blades
Leverancier: SWANN MORTON
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9275 9273 9272 9271
SWAN9275EA 66.8 EUR
SWAN9275 SWAN9273 SWAN9272 SWAN9271
Craft knife blades
Scalpels Scalpelmesjes
Surgical quality blades for use with handles No. 5A and No. 6A.
Verpakking: Supplied in anti-corrosion packaging.