Adjustable handles for brooms and squeegees
Leverancier: PERFEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8800S 53200-100
129-0128EA 24.5 EUR
129-0128 129-0127
Adjustable handles for brooms and squeegees
Bezems
These polymer handles are developed to meet the ergonomic and hygienic needs of controlled environments. They are widely accepted as a superior alternative to fixed length handles based on traditional materials, such as steel, aluminium and wood.
  • Comfortable and easy to use
  • Durable and lightweight
  • Handles will not corrode, oxidise, or rot
  • Compatible with gamma and ETO
  • Autoclavable up to 121 °C for 30 minutes
These handles are impervious to water, grease, detergents, sanitisers, and solvents. Grasp the upper handle and twist centre grip to adjust theMore Product Information