Chairs, Semi Stand-Up and Stools for Cleanrooms
Leverancier: Kango
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
7EK91GHLE01905 7EK91GHLE00905 7EK90GHLE00905 7EK90GBLN00905 7EK90GBLE00905 7EK91GBLE00905 7EK91GBLN00905 7EK90GHLE01905
BIEN7EK91GHLE01905EA 650 EUR
BIEN7EK91GHLE01905 BIEN7EK91GHLE00905 BIEN7EK90GHLE00905 BIEN7EK90GBLN00905 BIEN7EK90GBLE00905 BIEN7EK91GBLE00905 BIEN7EK91GBLN00905 BIEN7EK90GHLE01905
Chairs, Semi Stand-Up and Stools for Cleanrooms
Stoelen
Complete range of chairs, stools and semi stand-up seats developed for cleanrooms, in PU ESD or vinyl ESD.
  • Does not overheat or cause perspiration
  • Resists sharp tools, sparks, weak acids and hydrocarbons
  • Easily cleaned using common household products
  • Thickness of the foam: 4 cm
Different models and ranges to address all the working positions: Semi stand-up seats, comfort polyurethane chairs or Koncept chairs and stools
Auto-adaptative Koncept range in ESD Polyurethane:
A good working position is a dynamic posture, making it possible to move, because moving ensures good blood flow.
A seat tilted forward, split into two parts, based on silent-blocks forming a triangle helps to rebalance the body naturally and improve blood flow. The Koncept seat has great flexibility, which provides better distribution of pressure when moving.
The orientation of the seat also facilitates an upright posture and minimises spinal stress.
Dimensions of the seat (W×D): 410×400 mm
Dimensions of the lumbar support  (W×H) : 375x110mm (ref : 7EK)
Dimensions of the backrest (W×H): 340× 420 mm (ref : 7EP)
ESD Polyurethane comfort range:
Black ergonomic square seat and curved backrest for better lumbar support
Incorporated carry handle
Articulated backrest adjustable in height and depth (standard chairs)
Independently adjustable seat and backrest tilt with one fingertip lever thanks to an asynchronous mechanism (asynchronous chairs)
Dimensions of the seat (W×D): 410×390 mm
Dimensions of the backrest (W×H): 360× 220 mm
Gas lift column with shock absorber for height adjustment
5-branch scuff resistant reinforced base
Certificaten: ISO Class 3 certified (cleanrooms).