Safety blood lancets, VITREX® STERILANCE® Lite III
Leverancier: Vitrex Medical A/S
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
373213 374213 375213 376213 377213 378213 371213
MODU373213EA 18.6 EUR
MODU373213 MODU374213 MODU375213 MODU376213 MODU377213 MODU378213 MODU371213
Safety blood lancets, VITREX® STERILANCE® Lite III
Bloedlancetten
Safety lancets for blood glucose and other in vitro diagnostic tests.
  • Needle fully shielded before and after use
  • Pre-loaded pressure activated design
  • High quality, ultra sharp tri-bevel needle, high speed penetration
A variety of point configurations and penetration depths suit most capillary blood requirements.