Industriële handschoenen, Krynit 563
Leverancier: MAPA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
34563470 34563471 34563477 34563478 34563479
MAPA34563470EA 20.6 EUR
MAPA34563470 MAPA34563471 MAPA34563477 MAPA34563478 MAPA34563479
Industriële handschoenen, Krynit 563
Handschoenen
These seamless gloves made from HDPE fibres with nitrile coating on palm and fingertips provide protection for intricate handlings in dry and dirty environments.
  • Excellent life of duration thanks to an abrasion resistant coating
  • Excellent resistance to greases
  • Ventilated back for wearing comfort
  • Guaranteed without silicone and DMF
Typical applications include: Automotive/mechanical industry, pressing metal, handling metal sheets, mechanical maintenance, handling cardboard and paper, waste collection and handling, handling sheet metal.
Certificaten: EN 388 (4343B); Cat. II
ISO 13997: 5.8N (591 g)