Industriële handschoenen, Ultrane 551
Leverancier: MAPA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
34551030 34551031 34551036 34551037 34551038 34551039
MAPA34551030EA 267 EUR
MAPA34551030 MAPA34551031 MAPA34551036 MAPA34551037 MAPA34551038 MAPA34551039
Industriële handschoenen, Ultrane 551
Handschoenen
These seamless knitted gloves with PU foam coating on palm and fingertips provide excellent abrasion resistance.
  • Excellent tactile sensitivity due to reduced glove thickness
  • Absorption of perspiration ensured by the cellular structure of the PU coating
  • Ventilated back for additional wearing comfort
Typical applications include: Automotive/mechanical industry, sorting small parts, fitting small screws and fasteners, electronic component assembly (displays, LEDs, etc.), high-frequency antennae riveting, microprocessor handling, food industry, cosmetics and pharmaceutical industry, construction industry (metalworkers/locksmiths), installing and repairing locks, maintaining pipes, heating equipment, laying wood, panelling or moudling.
Certificaten: EN 388:2016 (4131X); Cat. II