Chemisch bestendige handschoenen, Ultranitril 491
Leverancier: MAPA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
34491266 34491267 34491268 34491260 34491269
MAPA34491266EA 297 EUR
MAPA34491266 MAPA34491267 MAPA34491268 MAPA34491260 MAPA34491269
Chemisch bestendige handschoenen, Ultranitril 491
Handschoenen
These anatomically designed gloves with flock lining provide good mechanical resistance and long-lasting chemical protection.
  • Dexterity and comfort thanks to the anatomical shape and the quality of the flock lining
  • Longer service life: Excellent mechanical resistance (abrasion, perforation)
  • Embossed texture for improved grip
Typical applications include: Automotive/mechanical industry, treating/degreasing metals with solvents, machining parts using cutting oil, maintenance, timber treatment and finishing, application of pesticides, cleaning of printing-press rollers, chemical industry, manufacture and application of paints and varnishes.
Certificaten: EN 388:2016 (3101X), EN ISO 374-1 Type A (AJKOPT), EN ISO 374-5 VIRUS; ISO 18889 G2; Cat. III